Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006719185.3 | XP_006719248.1 | 1 | 63 | 104 | |
| XM_011537729.2 | XP_011536031.1 | 1 | 62 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006719185.3, XM_011537729.2 transcripts.
Set Membership: |
|

YAF2
Genomic Map