Assay Details
| Gene Symbol: ST7-AS1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002330.1 | 1 | 1581 | 88 | |||
| GenBank mRNA | AF400039.1 | - | LIT2006 ST7OT1 |
1 | 1581 | 88 | |
More Information
Set Membership: |
|

ST7-AS1
Genomic Map