Assay Details
| Gene Symbol: MGC27345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046216.1 | 1 | 196 | 81 | ||
| Gene Symbol: RBM28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011516370.2 | XP_011514672.1 | 20 | 5731 | 81 | |
| XM_017012389.1 | XP_016867878.1 | 18 | 5507 | 81 | ||
| XM_017012390.1 | XP_016867879.1 | 17 | 5399 | 81 | ||
More Information
Set Membership: |
|

MGC27345
Genomic Map