Assay Details
| Gene Symbol: ANKRD30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519757.2 | XP_011518059.1 | 43 | 6139 | 86 | |
| XM_017016883.1 | XP_016872372.1 | 23 | 3968 | 86 | ||
More Information
Set Membership: |
|

ANKRD30A
Genomic Map