Assay Details
| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648555.1 | - | 1 | 149 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YAE1D1| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648555.1 | - | 1 | 149 | 138 | |
Set Membership: |
|