Assay Details
| Gene Symbol: URGCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077664.2 | NP_001071132.1 | 1 | 236 | 60 | |
| NM_001290076.1 | NP_001277005.1 | 1 | 236 | 60 | ||
| GenBank mRNA | AK302459.1 | - | 1 | 236 | 60 | |
| BG722670.1 | - | 1 | 214 | 60 | ||
| CD628752.1 | - | 1 | 229 | 60 | ||
| DC336174.1 | - | 1 | 236 | 60 | ||
| Gene Symbol: LOC107986791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012920.1 | XP_016868409.1 | 1 | 810 | 60 | |
| XM_017012921.1 | XP_016868410.1 | 1 | 810 | 60 | ||
| XM_017012922.1 | XP_016868411.1 | 1 | 810 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302459.1, NM_001077664.2, NM_001290076.1 transcripts.
Set Membership: |
|

URGCP
Genomic Map