Assay Details
| Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074039.1 | - | 5 | 2540 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STAG3L5P-PVRIG2P-PILRB| Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074039.1 | - | 5 | 2540 | 97 | |
Set Membership: |
|