Assay Details
| Gene Symbol: CUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202543.1 | NP_001189472.1 | 24 | 5801 | 107 | |
| NM_181552.3 | NP_853530.2 | 24 | 5679 | 107 | ||
| XM_005250150.2 | XP_005250207.1 | 24 | 6197 | 107 | ||
| XM_006715854.2 | XP_006715917.1 | 23 | 6134 | 107 | ||
| XM_011515823.2 | XP_011514125.1 | 24 | 5892 | 107 | ||
| XM_011515825.1 | XP_011514127.1 | 24 | 5460 | 107 | ||
| XM_017011760.1 | XP_016867249.1 | 23 | 5694 | 107 | ||
More Information
Set Membership: |
|

CUX1
Genomic Map