Assay Details
| Gene Symbol: DCAF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015420.6 | NP_056235.4 | 11 | 2223 | 85 | |
| GenBank mRNA | AK001693.1 | - | 10 | 4878 | 85 | |
| AK001874.1 | - | 11 | 1493 | 85 | ||
| BC026067.1 | - | 11 | 1548 | 85 | ||
| CA429438.1 | - | 1 | 85 | |||
More Information
Set Membership: |
|

DCAF13
Genomic Map