Assay Details
| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026974.1 | 1 | 2881 | 100 | ||
| GenBank mRNA | BX104600.1 | - | 1 | 108 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF252P-AS1| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026974.1 | 1 | 2881 | 100 | ||
| GenBank mRNA | BX104600.1 | - | 1 | 108 | 100 | |
Set Membership: |
|