Assay Details
| Gene Symbol: BMS1P20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX641141.1 | - | 5 | 2834 | 121 | |
| Gene Symbol: LOC101929959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017015379.1 | XP_016870868.1 | 7 | 1602 | 121 | |
More Information
Set Membership: |
|

BMS1P20
Genomic Map