Assay Details
| Gene Symbol: GPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004489.4 | NP_004480.1 | 5 | 496 | 98 | |
| GenBank mRNA | AK301877.1 | - | 6 | 508 | 98 | |
| BC013652.1 | - | 5 | 496 | 98 | ||
| BC103901.1 | - | 6 | 721 | 40005162 | 98 | |
| BC103902.1 | - | 5 | 1082 | 98 | ||
| BC103903.1 | - | 6 | 721 | 40005170 | 98 | |
| BC107738.1 | - | 6 | 531 | 5810755 | 98 | |
| BT006998.1 | - | 5 | 420 | 98 | ||
| CR541723.1 | - | 5 | 420 | 98 | ||
| CR541750.1 | - | 5 | 420 | 98 | ||
| U28963.1 | - | 6 | 510 | 98 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK096645.1 | - | 1858 | Chr. - - [-] on Build | 98 | ||
More Information
Set Membership: |
|

GPS2
Genomic Map