Assay Details
| Gene Symbol: AGTPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518420.2 | XP_011516722.1 | 1 | 3300 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011518420.2 transcripts.
Set Membership: |
|

AGTPBP1
Genomic Map