Assay Details
| Gene Symbol: PKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006717080.2 | XP_006717143.1 | 1 | 128 | 103 | |
| XM_017014649.1 | XP_016870138.1 | 1 | 130 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006717080.2, XM_017014649.1 transcripts.
Set Membership: |
|

PKN3
Genomic Map