Assay Details
| Gene Symbol: LINC00863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135292.1 | 4 - 5 | 1074 | 61 | ||
| GenBank mRNA | BC082979.1 | - | 4 - 5 | 879 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00863| Gene Symbol: LINC00863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135292.1 | 4 - 5 | 1074 | 61 | ||
| GenBank mRNA | BC082979.1 | - | 4 - 5 | 879 | 61 | |
Set Membership: |
|