Assay Details
| Gene Symbol: BLNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011539728.2 | XP_011538030.1 | 17 | 2539 | 113 | |
| XM_017016159.1 | XP_016871648.1 | 16 | 2471 | 113 | ||
More Information
Set Membership: |
|

BLNK
Genomic Map