Assay Details
| Gene Symbol: FTX | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028379.1 | 3 | 400 | 77 | |||
| GenBank mRNA | AK057701.1 | - | LIT1007 Ftx |
3 | 351 | 77 | |
| AK311345.1 | - | 3 | 400 | 77 | |||
More Information
Set Membership: |
|

FTX
Genomic Map