Assay Details
| Gene Symbol: LOC105373297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011531085.2 | XP_011529387.1 | 3 | 2766 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105373297| Gene Symbol: LOC105373297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011531085.2 | XP_011529387.1 | 3 | 2766 | 92 | |
Set Membership: |
|