Assay Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017029299.1 | XP_016884788.1 | 1 - 2 | 104 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017029299.1 transcripts.
Set Membership: |
|

USP51
Genomic Map