Assay Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005262126.3 | XP_005262183.1 | 5 | 3149 | 138 | |
| XM_006724639.3 | XP_006724702.1 | 4 | 2861 | 138 | ||
| XM_017029437.1 | XP_016884926.1 | 5 | 4208 | 138 | ||
| XM_017029438.1 | XP_016884927.1 | 3 | 2884 | 138 | ||
More Information
Set Membership: |
|

LPAR4
Genomic Map