Assay Details
| Gene Symbol: MID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005262062.4 | XP_005262119.1 | 1 | 427 | 110 | |
| XM_017029239.1 | XP_016884728.1 | 1 | 427 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005262062.4, XM_017029239.1 transcripts.
Set Membership: |
|

MID2
Genomic Map