Assay Details
| Gene Symbol: TARBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC150583.1 | - | 1 | 48 | 97 | |
| Gene Symbol: MAP3K12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538725.2 | XP_011537027.1 | 1 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538725.2 transcripts.
Set Membership: |
|

TARBP2
Genomic Map