Assay Details
| Gene Symbol: LOC105369781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135023.1 | 3 | 326 | 110 | ||
| GenBank mRNA | HY183314.1 | - | 1 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369781| Gene Symbol: LOC105369781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135023.1 | 3 | 326 | 110 | ||
| GenBank mRNA | HY183314.1 | - | 1 | 110 | ||
Set Membership: |
|