Assay Details
| Gene Symbol: PCDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173523.1 | NP_001166994.1 | 3 | 5291 | 205 | |
| NM_032457.3 | NP_115833.2 | 3 | 5267 | 205 | ||
| XM_005248163.3 | XP_005248220.1 | 4 | 5386 | 205 | ||
| XM_005248164.3 | XP_005248221.1 | 4 | 5362 | 205 | ||
| XM_005248166.3 | XP_005248223.1 | 2 | 5108 | 205 | ||
| XM_011513842.2 | XP_011512144.1 | 6 | 5450 | 205 | ||
| XM_011513843.2 | XP_011512145.1 | 5 | 5389 | 205 | ||
| XM_017008259.1 | XP_016863748.1 | 6 | 5426 | 205 | ||
| XM_017008260.1 | XP_016863749.1 | 5 | 5420 | 205 | ||
| XM_017008261.1 | XP_016863750.1 | 5 | 5417 | 205 | ||
| XM_017008262.1 | XP_016863751.1 | 5 | 5393 | 205 | ||
More Information
Set Membership: |
|

PCDH7
Genomic Map