Assay Details
| Gene Symbol: LOC440300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033738.1 | 8 | 4055 | 125 | ||
| GenBank mRNA | BX648930.1 | - | 8 | 4055 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440300| Gene Symbol: LOC440300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033738.1 | 8 | 4055 | 125 | ||
| GenBank mRNA | BX648930.1 | - | 8 | 4055 | 125 | |
Set Membership: |
|