Assay Details
Gene Symbol: C21orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011529623.2 | XP_011527925.1 | 1 | 2843 | 110 | |
XM_011529626.2 | XP_011527928.1 | 1 | 2843 | 110 | ||
XM_011529627.2 | XP_011527929.1 | 1 | 2843 | 110 | ||
XM_017028388.1 | XP_016883877.1 | 1 | 2843 | 110 | ||
XM_017028389.1 | XP_016883878.1 | 1 | 2843 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011529623.2, XM_011529626.2, XM_011529627.2, XM_017028388.1, XM_017028389.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
