Assay Details
| Gene Symbol: DGCR5 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110533.1 | 6 | 984 | 92 | |||
| GenBank mRNA | X91348.1 | - | LIT1227 DGCR5 |
6 | 93 | ||
More Information
Set Membership: |
|

DGCR5
Genomic Map