Assay Details
| Gene Symbol: DDTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011529816.2 | XP_011528118.1 | 1 | 311 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011529816.2 transcripts.
Set Membership: |
|

DDTL
Genomic Map