Assay Details
| Gene Symbol: MKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028889.1 | XP_016884378.1 | 1 | 873 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017028889.1 transcripts.
Set Membership: |
|

MKL1
Genomic Map