Assay Details
| Gene Symbol: TBC1D22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284303.1 | NP_001271232.1 | 11 | 3436 | 63 | |
| NM_001284304.1 | NP_001271233.1 | 14 | 3807 | 63 | ||
| NM_001284305.1 | NP_001271234.1 | 13 | 3747 | 63 | ||
| NM_014346.3 | NP_055161.1 | 13 | 3670 | 63 | ||
| NR_104292.1 | 14 | 4116 | 63 | |||
| XM_017028742.1 | XP_016884231.1 | 14 | 3762 | 63 | ||
| XM_017028744.1 | XP_016884233.1 | 14 | 3658 | 63 | ||
| XM_017028745.1 | XP_016884234.1 | 12 | 3489 | 63 | ||
| GenBank mRNA | AK000851.1 | - | 4 | 63 | ||
| DB339610.1 | - | 1 | 63 | |||
More Information
Set Membership: |
|

TBC1D22A
Genomic Map