Assay Details
| Gene Symbol: HEMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005265218.3 | XP_005265275.1 | 9 | 6779 | 118 | |
| XM_011533806.2 | XP_011532108.1 | 11 | 5704 | 118 | ||
| XM_011533807.1 | XP_011532109.1 | 11 | 5589 | 118 | ||
| XM_011533808.1 | XP_011532110.1 | 11 | 5375 | 118 | ||
| XM_011533809.2 | XP_011532111.1 | 12 | 5301 | 118 | ||
| XM_011533810.2 | XP_011532112.1 | 12 | 3344 | 118 | ||
| XM_011533812.2 | XP_011532114.1 | 9 | 5548 | 118 | ||
| XM_011533813.2 | XP_011532115.1 | 11 | 5742 | 118 | ||
| XM_011533814.2 | XP_011532116.1 | 11 | 5666 | 118 | ||
More Information
Set Membership: |
|

HEMK1
Genomic Map