Assay Details
| Gene Symbol: CAPN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319676.1 | NP_001306605.1 | 18 | 2046 | 95 | |
| NM_006615.2 | NP_006606.1 | 20 | 2235 | 95 | ||
| NM_016452.2 | NP_057536.1 | 19 | 2157 | 95 | ||
| XM_011544017.1 | XP_011542319.1 | 19 | 2160 | 95 | ||
| XM_011544018.1 | XP_011542320.1 | 18 | 2082 | 95 | ||
| XM_011544019.2 | XP_011542321.1 | 17 | 2082 | 95 | ||
| XM_011544020.1 | XP_011542322.1 | 19 | 1906 | 95 | ||
| XM_017000098.1 | XP_016855587.1 | 18 | 2076 | 95 | ||
| XM_017000099.1 | XP_016855588.1 | 20 | 1965 | 95 | ||
| GenBank mRNA | AB038463.1 | - | 19 | 2126 | 95 | |
| AF022799.1 | - | 20 | 2235 | 95 | ||
| BC027993.1 | - | 18 | 1997 | 5173512 | 95 | |
More Information
Set Membership: |
|

CAPN9
Genomic Map