Assay Details
| Gene Symbol: EBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009200.1 | XP_016864689.1 | 1 | 13581 | 61 | |
| XM_017009201.1 | XP_016864690.1 | 1 | 13581 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009200.1, XM_017009201.1 transcripts.
Set Membership: |
|

EBF1
Genomic Map