Assay Details
| Gene Symbol: TTBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011514948.2 | XP_011513250.1 | 1 | 1100 | 92 | |
| XM_017011368.1 | XP_016866857.1 | 1 | 1100 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011514948.2, XM_017011368.1 transcripts.
Set Membership: |
|

TTBK1
Genomic Map