Assay Details
Gene Symbol: LOC401286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_117091.1 | 3 | 2943 | 102 | ||
GenBank mRNA | AK123853.1 | - | 1 | 1485 | 102 | |
AK127120.1 | - | 3 | 2943 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
