Assay Details
| Gene Symbol: HEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010627.1 | XP_016866116.1 | 1 | 468 | 61 | |
| XM_017010628.1 | XP_016866117.1 | 2 | 166 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010627.1, XM_017010628.1 transcripts.
Set Membership: |
|

HEY2
Genomic Map