Assay Details
| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182762.3 | NP_877439.3 | 7 | 7626 | 80 | |
| GenBank mRNA | AL359055.1 | - | 2 | 1783 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MACC1| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182762.3 | NP_877439.3 | 7 | 7626 | 80 | |
| GenBank mRNA | AL359055.1 | - | 2 | 1783 | 80 | |
Set Membership: |
|