Assay Details
| Gene Symbol: TNS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011515479.2 | XP_011513781.1 | 1 | 1740 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011515479.2 transcripts.
Set Membership: |
|

TNS3
Genomic Map