Assay Details
| Gene Symbol: FZD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003505.1 | NP_003496.1 | 1 | 2732 | 97 | |
| GenBank mRNA | AB017363.1 | - | 1 | 2732 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FZD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003505.1 | NP_003496.1 | 1 | 2732 | 97 | |
| GenBank mRNA | AB017363.1 | - | 1 | 2732 | 97 | |
Set Membership: |
|