Assay Details
| Gene Symbol: GPAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178819.3 | NP_848934.1 | 13 | 2691 | 58 | |
| XM_011544392.2 | XP_011542694.1 | 14 | 2814 | 58 | ||
| XM_011544393.1 | XP_011542695.1 | 13 | 2703 | 58 | ||
| XM_011544394.1 | XP_011542696.1 | 13 | 2740 | 58 | ||
| XM_017013042.1 | XP_016868531.1 | 14 | 2841 | 58 | ||
| XM_017013043.1 | XP_016868532.1 | 14 | 2712 | 58 | ||
| GenBank mRNA | AL834255.1 | - | 9 | 1176 | 58 | |
| AY358670.1 | - | 12 | 1828 | 58 | ||
| AY513610.1 | - | 12 | 1911 | 58 | ||
| BC051377.1 | - | 12 | 1871 | 6649895 | 58 | |
| BC061884.1 | - | 12 | 1871 | 6577569 | 58 | |
More Information
Set Membership: |
|

GPAT4
Genomic Map