Assay Details
Gene Symbol: ZFHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024721.4 | NP_078997.4 | 10 | 6769 | 102 | |
XM_011517592.2 | XP_011515894.1 | 10 | 6529 | 102 | ||
XM_011517593.2 | XP_011515895.1 | 11 | 6523 | 102 | ||
XM_011517594.2 | XP_011515896.1 | 10 | 6534 | 102 | ||
XM_011517595.2 | XP_011515897.1 | 10 | 6603 | 102 | ||
XM_011517596.2 | XP_011515898.1 | 10 | 6752 | 102 | ||
XM_011517597.2 | XP_011515899.1 | 9 | 6713 | 102 | ||
XM_017013845.1 | XP_016869334.1 | 9 | 6635 | 102 | ||
GenBank mRNA | AB083343.2 | - | 12 | 6710 | 102 | |
AY260762.1 | - | 9 | 6291 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
