Assay Details
| Gene Symbol: VPS13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250801.4 | XP_005250858.1 | 1 | 1052 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250801.4 transcripts.
Set Membership: |
|

VPS13B
Genomic Map