Assay Details
| Gene Symbol: SMU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018225.2 | NP_060695.2 | 12 | 3397 | 81 | |
| XM_005251503.4 | XP_005251560.1 | 11 | 3233 | 81 | ||
| GenBank mRNA | AK001732.1 | - | 1 | 1466 | 81 | |
More Information
Set Membership: |
|

SMU1
Genomic Map