Assay Details
| Gene Symbol: UNC13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517681.2 | XP_011515983.1 | 2 | 4566 | 60 | |
| XM_011517682.2 | XP_011515984.1 | 2 | 4567 | 60 | ||
| XM_011517683.2 | XP_011515985.1 | 2 | 4562 | 60 | ||
More Information
Set Membership: |
|

UNC13B
Genomic Map