Assay Details
| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024496.1 | 2 | 2098 | 161 | ||
| NR_135010.1 | 3 | 1517 | 161 | |||
| GenBank mRNA | BC042539.1 | - | 2 | 2098 | 160 | |
| BC068605.1 | - | 3 | 1516 | 160 | ||
More Information
Set Membership: |
|

PTGER4P2-CDK2AP2P2
Genomic Map