Assay Details
| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005252674.4 | XP_005252731.1 | 1 | 119 | 85 | |
| XM_005252676.4 | XP_005252733.1 | 1 | 119 | 85 | ||
| XM_011539682.2 | XP_011537984.1 | 1 | 119 | 85 | ||
| XM_017016095.1 | XP_016871584.1 | 1 | 119 | 85 | ||
| XM_017016097.1 | XP_016871586.1 | 1 | 119 | 85 | ||
| XM_017016098.1 | XP_016871587.1 | 1 | 119 | 85 | ||
| XM_017016099.1 | XP_016871588.1 | 1 | 119 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005252674.4, XM_005252676.4, XM_011539682.2, XM_017016095.1, XM_017016097.1, XM_017016098.1, XM_017016099.1 transcripts.
Set Membership: |
|

JAKMIP3
Genomic Map