Assay Details
| Gene Symbol: ZFP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282515.1 | NP_001269444.1 | 4 | 950 | 173 | |
| NM_001282518.1 | NP_001269447.1 | 4 | 908 | 173 | ||
| NM_003408.2 | NP_003399.1 | 4 | 905 | 173 | ||
| GenBank mRNA | AF022158.1 | - | 4 | 830 | 173 | |
| AK301275.1 | - | 4 | 850 | 173 | ||
| BC126390.1 | - | 4 | 873 | 8992106 | 173 | |
More Information
Set Membership: |
|

ZFP37
Genomic Map