Assay Details
| Gene Symbol: MAN1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016219.4 | NP_057303.2 | 8 | 1269 | 88 | |
| NR_045720.1 | 8 | 1269 | 88 | |||
| NR_045721.1 | 9 | 1400 | 88 | |||
| XM_006716945.3 | XP_006717008.1 | 8 | 1334 | 88 | ||
| XM_017014239.1 | XP_016869728.1 | 9 | 1246 | 88 | ||
| GenBank mRNA | AF145732.1 | - | 8 | 1186 | 88 | |
| AF148509.1 | - | 8 | 1246 | 88 | ||
| AK074699.1 | - | 6 | 844 | 88 | ||
| AK130143.1 | - | 3 | 413 | 88 | ||
| AK298480.1 | - | 9 | 1246 | 88 | ||
| AK299043.1 | - | 8 | 1116 | 88 | ||
| AK309808.1 | - | 9 | 1246 | 88 | ||
| AK315797.1 | - | 8 | 1211 | 88 | ||
| AY358465.1 | - | 8 | 1218 | 88 | ||
| BC002953.1 | - | 8 | 1202 | 3533651 | 88 | |
| BC006079.1 | - | 8 | 1211 | 3959196 | 88 | |
| CR749534.1 | - | 9 | 1242 | 88 | ||
More Information
Set Membership: |
|

MAN1B1
Genomic Map