Assay Details
| Gene Symbol: RDX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260492.1 | NP_001247421.1 | 9 | 1198 | 122 | |
| NM_001260493.1 | NP_001247422.1 | 9 | 1198 | 122 | ||
| NM_001260494.1 | NP_001247423.1 | 7 | 827 | 122 | ||
| NM_002906.3 | NP_002897.1 | 9 | 1198 | 122 | ||
| GenBank mRNA | AK295075.1 | - | 6 | 543 | 122 | |
| AK312903.1 | - | 9 | 1100 | 122 | ||
| AK316061.1 | - | 7 | 729 | 122 | ||
| BC002626.2 | - | 9 | 1082 | 122 | ||
| BC020751.1 | - | 9 | 1098 | 122 | ||
| BC047109.1 | - | 9 | 1098 | 5284438 | 122 | |
| DQ916738.1 | - | 9 | 1198 | 122 | ||
| DQ916741.1 | - | 9 | 1198 | 122 | ||
| DQ916742.1 | - | 9 | 1198 | 122 | ||
| L02320.1 | - | 8 | 918 | 122 | ||
More Information
Set Membership: |
|

RDX
Genomic Map