Assay Details
| Gene Symbol: REEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011539501.2 | XP_011537803.1 | 6 | 9012 | 113 | |
| XM_017015896.1 | XP_016871385.1 | 7 | 7339 | 113 | ||
More Information
Set Membership: |
|

REEP3
Genomic Map